6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid ≥95%

Lieferant: Apollo Scientific

OR510102-250MG
APOSOR510102-250MGEA 163 EUR
APOSOR510102-250MG
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid ≥95%
6-Bromo-1H-pyrrolo[3,2-c]pyridine-3-carboxylic acid
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241,041 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09749989
CAS-Nummer: 1000341-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR