4-Bromo-5-nitro-1H-indole ≥97%
Lieferant: Apollo Scientific
OR510103-250MG
APOSOR510103-250MGEA
331
EUR
APOSOR510103-250MG
4-Bromo-5-nitro-1H-indole ≥97%
4-Bromo-5-nitro-1H-indole
Formel:
C₈H₅BrN₂O₂ Molecular Weight: 241,042 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
1934562-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...