6-Bromoimidazo[1,2-a]pyrazine-8-carboxylic acid ≥95%

Lieferant: Apollo Scientific

OR510104-250MG
APOSOR510104-250MGEA 271 EUR
APOSOR510104-250MG
6-Bromoimidazo[1,2-a]pyrazine-8-carboxylic acid ≥95%
6-Bromoimidazo[1,2-a]pyrazine-8-carboxylic acid
Formel: C₇H₄BrN₃O₂
Molecular Weight: 242,03 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13189932
CAS-Nummer: 1196154-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR