4-Chloro-2-methyl-5-(methylsulphonyl)benzoic acid ≥98%

Lieferant: Apollo Scientific

OR510106-250MG
APOSOR510106-250MGEA 0 EUR
APOSOR510106-250MG
4-Chloro-2-methyl-5-(methylsulphonyl)benzoic acid ≥98%
4-Chloro-2-methyl-5-(methylsulphonyl)benzoic acid
Formel: C₉H₉ClO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD21099591
CAS-Nummer: 176309-00-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR