2-(Hydroxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥98%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenemethanol

OR510107-250MG
APOSOR510107-250MGEA 125 EUR
APOSOR510107-250MG
2-(Hydroxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥98%
2-(Hydroxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formel: C₁₃H₁₉BO₄
Molecular Weight: 250,099 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18729969
CAS-Nummer: 760989-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR