Ethyl 5-amino-2-bromothiazole-4-carboxylate > 95%

Lieferant: Apollo Scientific

OR510108-250MG
APOSOR510108-250MGEA 197 EUR
APOSOR510108-250MG
Ethyl 5-amino-2-bromothiazole-4-carboxylate > 95%
Ethyl 5-amino-2-bromothiazole-4-carboxylate
Formel: C₆H₇BrN₂O₂S
Lagertemperatur: Gefrierschrank
MDL: MFCD27977252
CAS-Nummer: 1228281-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR