5-Bromo-4-nitrothiophene-2-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510109-250MG
APOSOR510109-250MGEA 102 EUR
APOSOR510109-250MG
5-Bromo-4-nitrothiophene-2-carboxylic acid ≥98%
5-Bromo-4-nitrothiophene-2-carboxylic acid
Formel: C₅H₂BrNO₄S
Molecular Weight: 252,043 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 89283-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR