2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole ≥97%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole

OR510110-250MG
APOSOR510110-250MGEA 91.5 EUR
APOSOR510110-250MG
2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole ≥97%
2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole
Formel: C₁₄H₁₈BNO₃
Molecular Weight: 259,109 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13181971
CAS-Nummer: 1408089-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR