4-Iod-3-nitroanilin ≥98%

Lieferant: Apollo Scientific

OR510111-1G
APOSOR510111-1GEA 36.4 EUR
APOSOR510111-1G
4-Iod-3-nitroanilin ≥98%
4-Iod-3-nitroanilin
Formel: C₆H₅IN₂O₂
Molecular Weight: 264,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08458843
CAS-Nummer: 105752-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR