7-Bromo-2-oxo-2H-chromene-3-carboxylic acid ≥96%

Lieferant: Apollo Scientific

OR510112-250MG
APOSOR510112-250MGEA 66.7 EUR
APOSOR510112-250MG
7-Bromo-2-oxo-2H-chromene-3-carboxylic acid ≥96%
7-Bromo-2-oxo-2H-chromene-3-carboxylic acid
Formel: C₁₀H₅BrO₄
Molecular Weight: 269,048 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1438410-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR