2-Iodo-6-methoxy-3-nitropyridine ≥98%

Lieferant: Apollo Scientific

OR510113-1G
APOSOR510113-1GEA 60.9 EUR
APOSOR510113-1G
2-Iodo-6-methoxy-3-nitropyridine ≥98%
2-Iodo-6-methoxy-3-nitropyridine
Formel: C₆H₅IN₂O₃
Molecular Weight: 280,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30241102
CAS-Nummer: 1600094-08-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR