Ethyl 2-amino-5-bromo-1H-indole-3-carboxylate ≥97%
Lieferant: Apollo Scientific
OR510114-250MG
APOSOR510114-250MGEA
65.4
EUR
APOSOR510114-250MG
Ethyl 2-amino-5-bromo-1H-indole-3-carboxylate ≥97%
Ethyl 2-amino-5-bromo-1H-indole-3-carboxylate
Formel:
C₁₁H₁₁BrN₂O₂ Molecular Weight: 283.121 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20226197 CAS-Nummer: 1242140-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...