Ethyl 6-bromo-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate > 97%

Lieferant: Apollo Scientific

OR510115-250MG
APOSOR510115-250MGEA 97.3 EUR
APOSOR510115-250MG
Ethyl 6-bromo-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate > 97%
Ethyl 6-bromo-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
Formel: C₁₁H₉BrN₂O₃
Molecular Weight: 297.105 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1446022-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR