tert-Butyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate ≥97%

Lieferant: Apollo Scientific

OR510116-250MG
APOSOR510116-250MGEA 74.1 EUR
APOSOR510116-250MG
tert-Butyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate ≥97%
tert-Butyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-1-carboxylate
Formel: C₁₂H₁₃BrN₂O₂
Lagertemperatur: Kühlschrank
CAS-Nummer: 1222809-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR