4,4'-Diphenyl-2,2'-bipyridine ≥97%
Lieferant: Apollo Scientific
OR510117-250MG
APOSOR510117-250MGEA
65.4
EUR
APOSOR510117-250MG
4,4'-Diphenyl-2,2'-bipyridine ≥97%
4,4'-Diphenyl-2,2'-bipyridine
Formel:
C₂₂H₁₆N₂ Molecular Weight: 308,376 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00006421 CAS-Nummer: 6153-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...