1-(Benzyloxy)-2-iodobenzene ≥98%

Lieferant: Apollo Scientific

OR510118-100MG
APOSOR510118-100MGEA 23.2 EUR
APOSOR510118-100MG
1-(Benzyloxy)-2-iodobenzene ≥98%
1-(Benzyloxy)-2-iodobenzene
Formel: C₁₃H₁₁IO
Molecular Weight: 310,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14648490
CAS-Nummer: 142523-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR