Ethyl 6-bromo-4,8-dichloroquinoline-3-carboxylate > 95%

Lieferant: Apollo Scientific

OR510119-1G
APOSOR510119-1GEA 82.8 EUR
APOSOR510119-1G
Ethyl 6-bromo-4,8-dichloroquinoline-3-carboxylate > 95%
Ethyl 6-bromo-4,8-dichloroquinoline-3-carboxylate
Formel: C₁₂H₈BrCl₂NO₂
Molecular Weight: 349,007 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD11522695
CAS-Nummer: 1019396-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR