N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 99%

Lieferant: Apollo Scientific

OR51012-1G OR51012-25G OR51012-5G
APOSOR51012-1GEA 21.9 EUR
APOSOR51012-1G APOSOR51012-25G APOSOR51012-5G
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 99%
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Formel: C₄₄H₃₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03093246
CAS-Nummer: 123847-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR