Ethyl 4-chloro-6-iodoquinoline-3-carboxylate > 97%

Lieferant: Apollo Scientific

OR510120-250MG
APOSOR510120-250MGEA 125 EUR
APOSOR510120-250MG
Ethyl 4-chloro-6-iodoquinoline-3-carboxylate > 97%
Ethyl 4-chloro-6-iodoquinoline-3-carboxylate
Formel: C₁₂H₉ClINO₂
Molecular Weight: 361,563 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02349007
CAS-Nummer: 206257-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR