6-Chloro-1-methyl-1H-indole-5-carboxylic acid ≥95%

Lieferant: Apollo Scientific

OR510121-1G
APOSOR510121-1GEA 352 EUR
APOSOR510121-1G
6-Chloro-1-methyl-1H-indole-5-carboxylic acid ≥95%
6-Chloro-1-methyl-1H-indole-5-carboxylic acid
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209,629 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08669342
CAS-Nummer: 431062-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR