3-Methyl-1H-pyrrole-2-carbaldehyde ≥98%

Lieferant: Apollo Scientific

OR510123-100MG
APOSOR510123-100MGEA 62.5 EUR
APOSOR510123-100MG
3-Methyl-1H-pyrrole-2-carbaldehyde ≥98%
3-Methyl-1H-pyrrole-2-carbaldehyde
Formel: C₆H₇NO
Molecular Weight: 109,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00030343
CAS-Nummer: 24014-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR