N3-Methylpyridine-3,4-diamine ≥98%

Lieferant: Apollo Scientific

OR510126-100MG
APOSOR510126-100MGEA 347 EUR
APOSOR510126-100MG
N3-Methylpyridine-3,4-diamine ≥98%
N3-Methylpyridine-3,4-diamine
Formel: C₆H₉N₃
Molecular Weight: 123,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384267
CAS-Nummer: 1796-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR