2,3-Dihydro-2-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
Lieferant: Apollo Scientific
OR510128-100MG
APOSOR510128-100MGEA
470
EUR
APOSOR510128-100MG
2,3-Dihydro-2-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
2,3-Dihydro-2-methyl-1H-pyrrolo[2,3-b]pyridine
Formel:
C₈H₁₀N₂ Molecular Weight: 134,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13175869 CAS-Nummer: 7546-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...