2,3-Dihydro-1H-pyrrolo[2,3-b]pyridin-4-ol ≥98%

Lieferant: Apollo Scientific

OR510129-100MG
APOSOR510129-100MGEA 464 EUR
APOSOR510129-100MG
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridin-4-ol ≥98%
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridin-4-ol
Formel: C₇H₈N₂O
Molecular Weight: 136,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22394516
CAS-Nummer: 1824145-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR