2,3-Dihydrofuro[2,3-b]pyridin-3-amine ≥98%

Lieferant: Apollo Scientific

OR510130-100MG
APOSOR510130-100MGEA 532 EUR
APOSOR510130-100MG
2,3-Dihydrofuro[2,3-b]pyridin-3-amine ≥98%
2,3-Dihydrofuro[2,3-b]pyridin-3-amine
Formel: C₇H₈N₂O
Molecular Weight: 136,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190275
CAS-Nummer: 870061-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR