(R)-2-methylpiperidine-2-carboxylic acid ≥98%
Lieferant: Apollo Scientific
OR510131-100MG
APOSOR510131-100MGEA
427
EUR
APOSOR510131-100MG
(R)-2-methylpiperidine-2-carboxylic acid ≥98%
(R)-2-methylpiperidine-2-carboxylic acid
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18089874 CAS-Nummer: 105141-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...