2-Hydroxy-4-(hydroxymethyl)benzaldehyde ≥98%

Lieferant: Apollo Scientific

OR510132-100MG
APOSOR510132-100MGEA 259 EUR
APOSOR510132-100MG
2-Hydroxy-4-(hydroxymethyl)benzaldehyde ≥98%
2-Hydroxy-4-(hydroxymethyl)benzaldehyde
Formel: C₈H₈O₃
Molecular Weight: 152,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24677929
CAS-Nummer: 156605-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR