2-Chloro-6-methylpyridine-4-carbonitrile ≥98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-6-methylisonicotinonitrile

OR510133-100MG
APOSOR510133-100MGEA 46.4 EUR
APOSOR510133-100MG
2-Chloro-6-methylpyridine-4-carbonitrile ≥98%
2-Chloro-6-methylpyridine-4-carbonitrile
Formel: C₇H₅ClN₂
Molecular Weight: 152,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658610
CAS-Nummer: 25462-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR