6-Chloro-4-methoxypyridin-2-amine ≥98%
Lieferant: Apollo Scientific
OR510134-100MG
APOSOR510134-100MGEA
52.2
EUR
APOSOR510134-100MG
6-Chloro-4-methoxypyridin-2-amine ≥98%
6-Chloro-4-methoxypyridin-2-amine
Formel:
C₆H₇ClN₂O Molecular Weight: 158,586 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18415424 CAS-Nummer: 439146-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...