Bicyclo[2.2.2]octan-1-amine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510135-100MG
APOSOR510135-100MGEA 352 EUR
APOSOR510135-100MG
Bicyclo[2.2.2]octan-1-amine hydrochloride ≥98%
Bicyclo[2.2.2]octan-1-amine hydrochloride
Formel: C₈H₁₅N·ClH
Molecular Weight: 161,672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28009517
CAS-Nummer: 1193-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR