5-Chloro-2-methyl-2H-pyrazolo[3,4-c]pyridine ≥98%

Lieferant: Apollo Scientific

OR510138-100MG
APOSOR510138-100MGEA 328 EUR
APOSOR510138-100MG
5-Chloro-2-methyl-2H-pyrazolo[3,4-c]pyridine ≥98%
5-Chloro-2-methyl-2H-pyrazolo[3,4-c]pyridine
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30830081
CAS-Nummer: 1951466-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR