4-Chlorothieno[2,3-b]pyridine ≥98%

Lieferant: Apollo Scientific

OR510139-100MG
APOSOR510139-100MGEA 30.6 EUR
APOSOR510139-100MG
4-Chlorothieno[2,3-b]pyridine ≥98%
4-Chlorothieno[2,3-b]pyridine
Formel: C₇H₄ClNS
Molecular Weight: 169,631 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858513
CAS-Nummer: 62226-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR