(3-Dibenzothiophen-4-ylphenyl)boronic acid 99%

Lieferant: Apollo Scientific

Synonyme: (3-(Dibenzo[b,d]thiophen-4-yl)phenyl)boronic acid

OR51014-5G
APOSOR51014-5GEA 115 EUR
APOSOR51014-5G
(3-Dibenzothiophen-4-ylphenyl)boronic acid 99%
(3-Dibenzothiophen-4-ylphenyl)boronic acid
Formel: C₁₈H₁₃BO₂S
Molecular Weight: 304.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200801
CAS-Nummer: 1307859-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR