5-(Chloromethyl)-3-cyclobutyl-1,2,4-oxadiazole ≥98%

Lieferant: Apollo Scientific

OR510140-100MG
APOSOR510140-100MGEA 241 EUR
APOSOR510140-100MG
5-(Chloromethyl)-3-cyclobutyl-1,2,4-oxadiazole ≥98%
5-(Chloromethyl)-3-cyclobutyl-1,2,4-oxadiazole
Formel: C₇H₉ClN₂O
Molecular Weight: 172,612 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15147267
CAS-Nummer: 1221278-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR