3-Azabicyclo[4.1.0]heptane-1-carboxylic acid hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510143-100MG
APOSOR510143-100MGEA 1320 EUR
APOSOR510143-100MG
3-Azabicyclo[4.1.0]heptane-1-carboxylic acid hydrochloride ≥98%
3-Azabicyclo[4.1.0]heptane-1-carboxylic acid hydrochloride
Formel: C₇H₁₁NO₂·ClH
Molecular Weight: 177,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858204
CAS-Nummer: 1181458-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR