7-Chloroisoquinolin-8-amine ≥98%

Lieferant: Apollo Scientific

OR510144-100MG
APOSOR510144-100MGEA 159 EUR
APOSOR510144-100MG
7-Chloroisoquinolin-8-amine ≥98%
7-Chloroisoquinolin-8-amine
Formel: C₉H₇ClN₂
Molecular Weight: 178,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18823041
CAS-Nummer: 55766-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR