2-Methylpiperidine-2-carboxylic acid hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510146-100MG
APOSOR510146-100MGEA 227 EUR
APOSOR510146-100MG
2-Methylpiperidine-2-carboxylic acid hydrochloride ≥98%
2-Methylpiperidine-2-carboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19103470
CAS-Nummer: 1094072-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR