(3-Chlorothiophen-2-yl)methanamine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510148-100MG
APOSOR510148-100MGEA 270 EUR
APOSOR510148-100MG
(3-Chlorothiophen-2-yl)methanamine hydrochloride ≥98%
(3-Chlorothiophen-2-yl)methanamine hydrochloride
Formel: C₅H₆ClNS·ClH
Molecular Weight: 184,087 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 643088-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR