2-Chloro-5-(2-hydroxyethyl)benzaldehyde ≥98%

Lieferant: Apollo Scientific

OR510149-1G
APOSOR510149-1GEA 1810 EUR
APOSOR510149-1G
2-Chloro-5-(2-hydroxyethyl)benzaldehyde ≥98%
2-Chloro-5-(2-hydroxyethyl)benzaldehyde
Formel: C₉H₉ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD24687256
CAS-Nummer: 1179337-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR