6-Benzyl-2-oxa-6-azaspiro[3.3]heptane ≥98%

Lieferant: Apollo Scientific

OR510151-100MG
APOSOR510151-100MGEA 1100 EUR
APOSOR510151-100MG
6-Benzyl-2-oxa-6-azaspiro[3.3]heptane ≥98%
6-Benzyl-2-oxa-6-azaspiro[3.3]heptane
Formel: C₁₂H₁₅NO
Molecular Weight: 189,254 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12407094
CAS-Nummer: 46246-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR