3-Bromo-6-methylpicolinaldehyde ≥98%

Lieferant: Apollo Scientific

OR510152-100MG
APOSOR510152-100MGEA 322 EUR
APOSOR510152-100MG
3-Bromo-6-methylpicolinaldehyde ≥98%
3-Bromo-6-methylpicolinaldehyde
Formel: C₇H₆BrNO
Molecular Weight: 200,033 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189310
CAS-Nummer: 1060810-14-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR