Methyl 4-bromofuran-3-carboxylate 95%

Lieferant: Apollo Scientific

OR510154-100MG
APOSOR510154-100MGEA 612 EUR
APOSOR510154-100MG
Methyl 4-bromofuran-3-carboxylate 95%
Methyl 4-bromofuran-3-carboxylate
Formel: C₆H₅BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD32640807
CAS-Nummer: 2069255-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR