5-Bromo-2-isopropylthiazole ≥98%

Lieferant: Apollo Scientific

OR510155-100MG
APOSOR510155-100MGEA 296 EUR
APOSOR510155-100MG
5-Bromo-2-isopropylthiazole ≥98%
5-Bromo-2-isopropylthiazole
Formel: C₆H₈BrNS
Molecular Weight: 206,103 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1086382-46-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR