4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%

Lieferant: Apollo Scientific

OR510156-100MG
APOSOR510156-100MGEA 644 EUR
APOSOR510156-100MG
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine ≥98%
4-Bromo-6-methyl-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09880145
CAS-Nummer: 1000340-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR