2,6-Dichloro-4-(dimethylamino)nicotinaldehyde ≥98%

Lieferant: Apollo Scientific

OR510157-100MG
APOSOR510157-100MGEA 444 EUR
APOSOR510157-100MG
2,6-Dichloro-4-(dimethylamino)nicotinaldehyde ≥98%
2,6-Dichloro-4-(dimethylamino)nicotinaldehyde
Formel: C₈H₈Cl₂N₂O
Molecular Weight: 219,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24873202
CAS-Nummer: 1823371-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR