7-Bromonaphthalen-1-amine ≥98%
Lieferant: Apollo Scientific
OR510159-100MG
APOSOR510159-100MGEA
55.1
EUR
APOSOR510159-100MG
7-Bromonaphthalen-1-amine ≥98%
7-Bromonaphthalen-1-amine
Formel:
C₁₀H₈BrN Molecular Weight: 222,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17012507 CAS-Nummer: 136924-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...