Fluoranthene-3-boronic acid 98%

Lieferant: Apollo Scientific

Synonyme: Fluoranthen-3-ylboronic acid

OR51016-5G
APOSOR51016-5GEA 186 EUR
APOSOR51016-5G
Fluoranthene-3-boronic acid 98%
Fluoranthene-3-boronic acid
Formel: C₁₆H₁₁BO₂
Molecular Weight: 246.0683 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16660041
CAS-Nummer: 359012-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR