1-Bromo-2,6-naphthyridin-3-amine ≥98%

Lieferant: Apollo Scientific

OR510160-250MG
APOSOR510160-250MGEA 1100 EUR
APOSOR510160-250MG
1-Bromo-2,6-naphthyridin-3-amine ≥98%
1-Bromo-2,6-naphthyridin-3-amine
Formel: C₈H₆BrN₃
Molecular Weight: 224,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00160697
CAS-Nummer: 3611-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR