(7-Chloronaphthalen-1-yl)methanamine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510161-100MG
APOSOR510161-100MGEA 655 EUR
APOSOR510161-100MG
(7-Chloronaphthalen-1-yl)methanamine hydrochloride ≥98%
(7-Chloronaphthalen-1-yl)methanamine hydrochloride
Formel: C₁₁H₁₀ClN·ClH
Molecular Weight: 228,118 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31617675
CAS-Nummer: 1093651-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR