5-Bromo-7-chloro-1H-pyrazolo[3,4-c]pyridine ≥98%

Lieferant: Apollo Scientific

OR510162-100MG
APOSOR510162-100MGEA 447 EUR
APOSOR510162-100MG
5-Bromo-7-chloro-1H-pyrazolo[3,4-c]pyridine ≥98%
5-Bromo-7-chloro-1H-pyrazolo[3,4-c]pyridine
Formel: C₆H₃BrClN₃
Molecular Weight: 232,465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21603740
CAS-Nummer: 1416374-15-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR